accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-10-1

IUPAC Name: 2,6-dimethoxyphenol
Synonyms: 2,6-dimethoxyphenol syringol 1,3-di-o-methylpyrogallol pyrogallol 1,3-dimethyl ether 1,3-dimethyl pyrogallate pyrogallol dimethylether aldrich 2,6-dwumetoksyfenol
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: KLIDCXVFHGNTTM-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1O
Percent Purity 
  • (1)
  • (3)
  • (7)
Boiling Point 
  • (5)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (4)
  • (4)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (3)
  • (7)
  • (5)
  • (2)
  • (3)
  • (2)
  • (8)
  • (4)
  • (4)
  • (2)
  • (3)
  • (3)
  • (1)