CAS RN 91-17-8
Decahydronaphthalene, 98%, mixture of cis and trans
CAS: 91-17-8 Molecular Formula: C10H18 Molecular Weight (g/mol): 138.25 MDL Number: MFCD00004130 InChI Key: NNBZCPXTIHJBJL-UHFFFAOYSA-N Synonym: decalin,decahydronaphthalene,cis-decahydronaphthalene,trans-decahydronaphthalene,cis-decalin,dekalin,naphthalene, decahydro,perhydronaphthalene,trans-decalin,naphthalane PubChem CID: 7044 ChEBI: CHEBI:38853 IUPAC Name: 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene SMILES: C1CCC2CCCCC2C1
Decahydronaphthalene, cis + trans, 98%
CAS: 91-17-8 Molecular Formula: C10H18 Molecular Weight (g/mol): 138.254 MDL Number: MFCD00004130 InChI Key: NNBZCPXTIHJBJL-UHFFFAOYSA-N Synonym: decalin,decahydronaphthalene,cis-decahydronaphthalene,trans-decahydronaphthalene,cis-decalin,dekalin,naphthalene, decahydro,perhydronaphthalene,trans-decalin,naphthalane PubChem CID: 7044 ChEBI: CHEBI:38853 IUPAC Name: 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene SMILES: C1CCC2CCCCC2C1
Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™
CAS: 91-17-8 Molecular Formula: C10H18 Molecular Weight (g/mol): 138.25 InChI Key: NNBZCPXTIHJBJL-UHFFFAOYSA-N Synonym: decalin,decahydronaphthalene,cis-decahydronaphthalene,trans-decahydronaphthalene,cis-decalin,dekalin,naphthalene, decahydro,perhydronaphthalene,trans-decalin,naphthalane PubChem CID: 7044 ChEBI: CHEBI:38853 IUPAC Name: 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene SMILES: C1CCC2CCCCC2C1
Decahydronaphthalene (cis- and trans- mixture) 99.0+%, TCI America™
CAS: 91-17-8 Molecular Formula: C10H18 Molecular Weight (g/mol): 138.254 MDL Number: MFCD00004130 InChI Key: NNBZCPXTIHJBJL-UHFFFAOYSA-N Synonym: decalin,decahydronaphthalene,cis-decahydronaphthalene,trans-decahydronaphthalene,cis-decalin,dekalin,naphthalene, decahydro,perhydronaphthalene,trans-decalin,naphthalane PubChem CID: 7044 ChEBI: CHEBI:38853 IUPAC Name: 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene SMILES: C1CCC2CCCCC2C1
Decahydronaphthalene, Mixture of cis + trans, 98%, Reagent Grade, Decalin™, Solstice
CAS: 91-17-8 Molecular Formula: C10H18 Molecular Weight (g/mol): 138.254 MDL Number: MFCD00004130 InChI Key: NNBZCPXTIHJBJL-UHFFFAOYSA-N Synonym: decalin,decahydronaphthalene,cis-decahydronaphthalene,trans-decahydronaphthalene,cis-decalin,dekalin,naphthalene, decahydro,perhydronaphthalene,trans-decalin,naphthalane PubChem CID: 7044 ChEBI: CHEBI:38853 IUPAC Name: 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene SMILES: C1CCC2CCCCC2C1
Decahydronaphthalene (cis- and trans- mixture) 97.0+%, TCI America™
CAS: 91-17-8 Molecular Formula: C10H18 Molecular Weight (g/mol): 138.254 MDL Number: MFCD00004130 InChI Key: NNBZCPXTIHJBJL-UHFFFAOYSA-N Synonym: decalin,decahydronaphthalene,cis-decahydronaphthalene,trans-decahydronaphthalene,cis-decalin,dekalin,naphthalene, decahydro,perhydronaphthalene,trans-decalin,naphthalane PubChem CID: 7044 ChEBI: CHEBI:38853 IUPAC Name: 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene SMILES: C1CCC2CCCCC2C1
Decahydronaphthalene (Laboratory), Fisher Chemical™
CAS: 91-17-8 Molecular Formula: C10H18 Molecular Weight (g/mol): 138.254 MDL Number: MFCD00004130 InChI Key: NNBZCPXTIHJBJL-UHFFFAOYSA-N Synonym: decalin,decahydronaphthalene,cis-decahydronaphthalene,trans-decahydronaphthalene,cis-decalin,dekalin,naphthalene, decahydro,perhydronaphthalene,trans-decalin,naphthalane PubChem CID: 7044 ChEBI: CHEBI:38853 IUPAC Name: 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene SMILES: C1CCC2CCCCC2C1