Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decahydronaphthalene, Mixture of cis + trans, 98%, Reagent Grade, Decalin™, Solstice

Catalog No. 60047951 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Aluminum bottle
Glass bottle
Stainless steel can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-951 4 L Aluminum bottle
60-047-950 20 L Stainless steel can
60-047-949 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-047-951 Supplier Solstice Supplier No. D2514L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00004130
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Boiling Range 189° - 191°C
Melting Point -125°C
Density 0.896g/mL at 25°C
Explosion Limit 0.7 to 4.9%, 100°F
Flash Point 57°C (134.6°F)
Ignition Point 482°F
Packaging Aluminum bottle
Quantity 4 L
UN Number UN1147
Beilstein 878165
Vapor Density 4.76 (vs air)
Vapor Pressure 42mmHg (92°C), 741mmHg (188°C)
Formula Weight 138.25g/mol
Percent Purity 0.98
Chemical Name or Material Decahydronaphthalene, mixture of cis plus trans
Show More Show Less
Hazard Statement P261-P273-P280-P301 + P310-P305 + P351 + P338-P310
DOTInformation class 3 - PG 3 - Decahydronaphthalene
RTECSNumber 202-046-9
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.