CAS RN 91-20-3
Naphthalene, 99%
CAS: 91-20-3 Molecular Formula: C10H8 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00001742 InChI Key: UFWIBTONFRDIAS-UHFFFAOYSA-N Synonym: naphthalin,naphthene,camphor tar,tar camphor,white tar,albocarbon,naphthaline,moth flakes,moth balls,naphtalene PubChem CID: 931 ChEBI: CHEBI:16482 IUPAC Name: naphthalene SMILES: C1=CC2=CC=CC=C2C=C1
Naphthalene, 99+%, scintillation grade
CAS: 91-20-3 Molecular Formula: C10H8 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00001742 InChI Key: UFWIBTONFRDIAS-UHFFFAOYSA-N PubChem CID: 931 ChEBI: CHEBI:16482 IUPAC Name: naphthalene SMILES: C1=CC2=CC=CC=C2C=C1
Naphthalene, 99%
CAS: 91-20-3 Molecular Formula: C10H8 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00001742 InChI Key: UFWIBTONFRDIAS-UHFFFAOYSA-N Synonym: naphthalin,naphthene,camphor tar,tar camphor,white tar,albocarbon,naphthaline,moth flakes,moth balls,naphtalene PubChem CID: 931 ChEBI: CHEBI:16482 IUPAC Name: naphthalene SMILES: C1=CC2=CC=CC=C2C=C1
Naphthalene, 99+%
CAS: 91-20-3 Molecular Formula: C10H8 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00001742 InChI Key: UFWIBTONFRDIAS-UHFFFAOYSA-N Synonym: naphthalin,naphthene,camphor tar,tar camphor,white tar,albocarbon,naphthaline,moth flakes,moth balls,naphtalene PubChem CID: 931 ChEBI: CHEBI:16482 IUPAC Name: naphthalene SMILES: C1=CC2=CC=CC=C2C=C1
Melting point Standard 79-81C, MilliporeSigma™ Supelco™
Melting point standard 79-81C (Naphthalene) is a hydrocarbon which is nonpolar in nature. It is soluble in toluene. It can be crystallized from ether.
Naphthalene 98.0+%, TCI America™
CAS: 91-20-3 Molecular Formula: C10H8 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00001742 InChI Key: UFWIBTONFRDIAS-UHFFFAOYSA-N Synonym: naphthalin,naphthene,camphor tar,tar camphor,white tar,albocarbon,naphthaline,moth flakes,moth balls,naphtalene PubChem CID: 931 ChEBI: CHEBI:16482 IUPAC Name: naphthalene SMILES: C1=CC2=CC=CC=C2C=C1
Sigma Aldrich Naphthalene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Agilent Technologies Naphthalene Solution, 5000μg/mL in Methanol, Ultra Scientific
CAS: 91-20-3 Molecular Formula: C10H8 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00001742 InChI Key: UFWIBTONFRDIAS-UHFFFAOYSA-N PubChem CID: 931 ChEBI: CHEBI:16482 IUPAC Name: naphthalene SMILES: C1=CC2=CC=CC=C2C=C1
Sigma Aldrich Naphthalene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Naphthalene, Scintillation Grade, ≥99%, MP Biomedicals™
CAS: 91-20-3 Molecular Formula: C10H8 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00001742 InChI Key: UFWIBTONFRDIAS-UHFFFAOYSA-N Synonym: naphthalin,naphthene,camphor tar,tar camphor,white tar,albocarbon,naphthaline,moth flakes,moth balls,naphtalene PubChem CID: 931 ChEBI: CHEBI:16482 IUPAC Name: naphthalene SMILES: C1=CC2=CC=CC=C2C=C1
Fisher Science Education™ Naphthalene
CAS: 91-20-3 Molecular Formula: C10H8 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00001742 InChI Key: UFWIBTONFRDIAS-UHFFFAOYSA-N Synonym: naphthalin,naphthene,camphor tar,tar camphor,white tar,albocarbon,naphthaline,moth flakes,moth balls,naphtalene PubChem CID: 931 ChEBI: CHEBI:16482 IUPAC Name: naphthalene SMILES: C1=CC2=CC=CC=C2C=C1