Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthalene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017862 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7862 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7862 Supplier MilliporeSigma Supelco Supplier No. PHR12751G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standard certified reference material for quality control applications.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naphthalene
CAS 91-20-3
Ignition Point 978°F
Vapor Pressure 0.03 mmHg (25°C)
Vapor Density 4.4 (vs. air)
Flash Point 80.0°C - Closed Cup
Physical Form Neat
Linear Formula C10H8
Technique HPLC
MDL Number MFCD00001742
Certifications/Compliance ISO/IEC 17025
Melting Point 80°C to 82°C
Boiling Point 218°C
Synonym Naphthalene
Type Certified Reference Material
Industry Type Pharmaceutical
Molecular Weight (g/mol) 128.17
Quantity 1 g
Formula Weight 128.17
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.