accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOOH3C

CAS RN 91-52-1

IUPAC Name: 2,4-dimethoxybenzoic acid
Synonyms: 2,4-dimethoxybenzoic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: GPVDHNVGGIAOQT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(O)=O)C(OC)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)