accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-67-8

IUPAC Name: N,N-diethyl-3-methylaniline
Synonyms: N,N-diethyl-3-methylaniline
Molecular Weight (g/mol): 163.26
Molecular Formula: C11H17N
InChi Key: CIPVVROJHKLHJI-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=CC(C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results