Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-m-toluidine 99.0+%, TCI America™
SDP

Catalog No. D38685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38685G 5 g
D38681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38685G Supplier TCI America Supplier No. D38685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91-67-8
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.26
MDL Number MFCD00035795
InChI Key CIPVVROJHKLHJI-UHFFFAOYSA-N
Synonym n,n-diethyl-m-toluidine, benzenamine, n,n-diethyl-3-methyl, 3-diethylamino toluene, 3-n,n-diethylamino toluene, m-methyl-n,n-diethylaniline, 1-diethylamino-3-methylbenzene, 3-diethylamino-1-methylbenzene, n,n-diethyl-m-toluidinium ion, m-toluidine, n,n-diethyl, meta-methyl diethylamino benzene
PubChem CID 66679
IUPAC Name N,N-diethyl-3-methylaniline
SMILES CCN(CC)C1=CC=CC(C)=C1

Specifications

Color Yellow
Boiling Point 107°C
Quantity 5 g
Formula Weight 163.26
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-m-toluidine [for Biochemical Research]
EINECSNumber (3)-0191
RTECSNumber CX9869375
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.