accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-77-0

IUPAC Name: N2,N2-bis(prop-2-en-1-yl)-1,3,5-triazine-2,4,6-triamine
Synonyms: N2,N2-bis(prop-2-en-1-yl)-1,3,5-triazine-2,4,6-triamine
Molecular Weight (g/mol): 206.25
Molecular Formula: C9H14N6
InChi Key: ROHTVIURAJBDES-UHFFFAOYSA-N
SMILES: NC1=NC(=NC(N)=N1)N(CC=C)CC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results