Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-6-diallylamino-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. D1075500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1075500G 500 g
D107525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1075500G Supplier TCI America Supplier No. D1075500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91-77-0
Molecular Formula C9H14N6
Molecular Weight (g/mol) 206.253
MDL Number MFCD00047347
InChI Key ROHTVIURAJBDES-UHFFFAOYSA-N
Synonym N2,N2-Diallylmelamine
PubChem CID 66682
IUPAC Name 2-N,2-N-bis(prop-2-enyl)-1,3,5-triazine-2,4,6-triamine
SMILES C=CCN(CC=C)C1=NC(=NC(=N1)N)N

Specifications

Melting Point 145°C
Color White
Quantity 500 g
Formula Weight 206.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-diallylamino-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.