accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91-88-3

IUPAC Name: 2-[ethyl(3-methylphenyl)amino]ethan-1-ol
Synonyms: 2-[ethyl(3-methylphenyl)amino]ethanol
Molecular Weight (g/mol): 179.26
Molecular Formula: C11H17NO
InChi Key: KRNUKKZDGDAWBF-UHFFFAOYSA-N
SMILES: CCN(CCO)C1=CC=CC(C)=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)