Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(N-Ethyl-N-m-toluidino)ethanol

Catalog No. 128325100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-832-5100G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-832-5100G Supplier Sigma Aldrich Supplier No. 128325100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(N-Ethyl-N-m-toluidino)ethanol

Specifications

CAS 91-88-3
Density 1.019 g/mL (at 25°C (literature))
Boiling Point 114°C to 115°C (1 mmHg)
Molecular Formula C11H17NO
Refractive Index n20/D 1.555 (literature)
Linear Formula CH3C6H4N(C2H5)CH2CH2OH
MDL Number MFCD00002849
Quantity 100 mg
Synonym N-Ethyl-N-(2-hydroxyethyl)-m-toluidine
Molecular Weight (g/mol) 179.26
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.