accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 910036-98-5

IUPAC Name: 2-(oxan-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-(oxan-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 305.18
Molecular Formula: C16H24BNO4
InChi Key: KGCJDMWHIVJQGF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results