accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 910037-07-9

IUPAC Name: methyl({[4-(4-methyl-1,4-diazepan-1-yl)phenyl]methyl})amine
Synonyms: methyl({[4-(4-methyl-1,4-diazepan-1-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 233.36
Molecular Formula: C14H23N3
InChi Key: NOEOYDRQGMQWID-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)N1CCCN(C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results