accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 910251-35-3

IUPAC Name: potassium [(acetyloxy)methyl]trifluoroboranuide
Synonyms: potassium [(acetyloxy)methyl]trifluoroboranuide
Molecular Weight (g/mol): 179.97
Molecular Formula: C3H5BF3KO2
InChi Key: CVJAGVAZVCBBFZ-UHFFFAOYSA-N
SMILES: [K+].CC(=O)OC[B-](F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results