Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium (Acetoxymethyl)trifluoroborate 96.0+%, TCI America™
SDP

Catalog No. P24941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P24941G Supplier TCI America Supplier No. P24941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 910251-35-3
Molecular Formula C3H5BF3KO2
Molecular Weight (g/mol) 179.97
MDL Number MFCD28023894
InChI Key CVJAGVAZVCBBFZ-UHFFFAOYSA-N
PubChem CID 88542032
IUPAC Name potassium [(acetyloxy)methyl]trifluoroboranuide
SMILES [K+].CC(=O)OC[B-](F)(F)F

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 g
Formula Weight 179.97
Percent Purity ≥97.0% (T)
Chemical Name or Material Potassium (Acetoxymethyl)trifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.