CAS RN 91053-39-3
Celite(R), 521, Spectrum™ Chemical
CAS: 91053-39-3 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N IUPAC Name: silanedione SMILES: O=[Si]=O
Filter aid, Celite Standard Super-cel
Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 MDL Number: MFCD00132803 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Residue Sample 1, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Residue, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Particulate Matter on Filter Paper, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Residue - WP, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.