CAS RN 91076-93-6
CAS RN 91076-93-6
IUPAC Name:
methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
Synonyms:
methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
Molecular Weight (g/mol):
267.73
Molecular Formula:
C12H10ClNO2S
InChi Key:
MELGGDOYSMRBGA-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(N)C=C(S1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Methyle3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate, 97%, Thermo Scientific™
CAS: 91076-93-6 Molecular Formula: C12H10ClNO2S Molecular Weight (g/mol): 267.727 MDL Number: MFCD00052591 InChI Key: MELGGDOYSMRBGA-UHFFFAOYSA-N PubChem CID: 2777492 IUPAC Name: methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate SMILES: COC(=O)C1=C(C=C(S1)C2=CC=C(C=C2)Cl)N