Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog No. GK00598EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
GK00598EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GK00598EA Supplier Thermo Scientific Chemicals Supplier No. GK00598EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91076-93-6
Molecular Formula C12H10ClNO2S
Molecular Weight (g/mol) 267.727
MDL Number MFCD00052591
InChI Key MELGGDOYSMRBGA-UHFFFAOYSA-N
PubChem CID 2777492
IUPAC Name methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
SMILES COC(=O)C1=C(C=C(S1)C2=CC=C(C=C2)Cl)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym methyl 3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl3-amino-5-4-chlorophenyl thiophene-2-carboxylate, methyl-3-amino-5-4-chlorophenyl thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-amino-5-4-chlorophenyl-, methyl ester, 3-amino-5-4-chlorophenyl thiophene-2-carboxylic acid methyl ester, pubchem8387, maybridge1_003891, methyl 3-amino-5-4-chlorophenyl thiophene-2carboxylate, methyl 3-amino-5-4-chlorophenyl-2-thiophenecarboxylate, methyl 3-amino-5-4-chlorophenyl-thiophene-2-carboxylate
Formula Weight 267.74
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.