accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 910805-49-1

IUPAC Name: 5-benzyl-12-phenyl-7-(phenylsulfanyl)-3,8,11,13-tetraoxa-5-azatricyclo[7.4.0.0²,⁶]tridecan-4-one
Synonyms: 5-benzyl-12-phenyl-7-(phenylsulfanyl)-3,8,11,13-tetraoxa-5-azatricyclo[7.4.0.0²,⁶]tridecan-4-one
Molecular Weight (g/mol): 475.56
Molecular Formula: C27H25NO5S
InChi Key: IPBJFGDRNYVDGL-UHFFFAOYNA-N
SMILES: O=C1OC2C(C(OC3COC(OC23)C2=CC=CC=C2)SC2=CC=CC=C2)N1CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results