Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl N-Benzyl-2-amino-4,6-O-benzylidene-2-N,3-O-carbonyl-2-deoxy-1-thio-beta-D-glucopyranoside 97.0+%, TCI America™
SDP

Catalog No. P17621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P17621G Supplier TCI America Supplier No. P17621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 910805-49-1
Molecular Formula C27H25NO5S
Molecular Weight (g/mol) 475.559
InChI Key IPBJFGDRNYVDGL-JVIQZWCRSA-N
PubChem CID 101517088
IUPAC Name (2R,4aR,6S,6aR,9aR,9bS)-7-benzyl-2-phenyl-6-phenylsulfanyl-4,4a,6,6a,9a,9b-hexahydro-[1,3]dioxino[4,5]pyrano[1,2-b][1,3]oxazol-8-one
SMILES C1C2C(C3C(C(O2)SC4=CC=CC=C4)N(C(=O)O3)CC5=CC=CC=C5)OC(O1)C6=CC=CC=C6

Specifications

Color White
Quantity 1 g
Formula Weight 475.56
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl N-Benzyl-2-amino-4,6-O-benzylidene-2-N,3-O-carbonyl-2-deoxy-1-thio-beta-D-glucopyranoside
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.