accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH(R)SNOOOHO

CAS RN 91088-55-0

IUPAC Name: (2R)-2-acetamido-3-[(4-nitrophenyl)sulfanyl]propanoic acid
Synonyms: (2R)-2-acetamido-3-[(4-nitrophenyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 284.29
Molecular Formula: C11H12N2O5S
InChi Key: LOBKSYPZBSVLLE-JTQLQIEISA-N
SMILES: CC(=O)N[C@@H](CSC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results