Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-S-(4-nitrophenyl)-L-cysteine 98.0+%, TCI America™
SDP

Catalog No. A1351100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1351100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1351100MG Supplier TCI America Supplier No. A1351100MG

Chemical Identifiers

CAS 91088-55-0
Molecular Formula C11H12N2O5S
Molecular Weight (g/mol) 284.286
MDL Number MFCD00671484
InChI Key LOBKSYPZBSVLLE-JTQLQIEISA-N
Synonym (R)-2-Acetamido-3-(4-nitrophenylthio)propionic Acid, L-4-Nitrophenylmercapturic Acid
PubChem CID 185069
IUPAC Name (2R)-2-acetamido-3-(4-nitrophenyl)sulfanylpropanoic acid
SMILES CC(=O)NC(CSC1=CC=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 158°C
Color Yellow
Quantity 100 mg
Formula Weight 284.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-S-(4-nitrophenyl)-L-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.