accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CNHOOCH3CH3H3C

CAS RN 91103-47-8

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: methyl 2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: GJDICGOCZGRDFM-UHFFFAOYNA-N
SMILES: COC(=O)C(C)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)