accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOONOOH3CCH3H3C

CAS RN 91229-91-3

IUPAC Name: 1,2-di-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate
Synonyms: 1,2-di-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 285.34
Molecular Formula: C14H23NO5
InChi Key: INVKHBRFFCQICU-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results