Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl N-(tert-Butoxycarbonyl)-L-pyroglutamate 98.0+%, TCI America™
SDP

Catalog No. B52165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52165G 5 g
B52161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52165G Supplier TCI America Supplier No. B52165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 91229-91-3
Molecular Formula C14H23NO5
Molecular Weight (g/mol) 285.34
MDL Number MFCD03094770
InChI Key INVKHBRFFCQICU-VIFPVBQESA-N
Synonym s-di-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate, s-n-boc-2-pyrrolidone-5-carboxylic acid tert-butyl ester, s-n-boc-2-pyrrolidone-5-carboxylic acid t-butyl ester, s-n-boc-pyroglutamic acid tert-butyl ester, s-n-alpha-t-butyloxycarbonyl-pyroglutamic acid t-butyl ester, di-tert-butyl s-5-oxopyrrolidine-1,2-dicarboxylate, 1,2-di-tert-butyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, s-n-boc-pyroglutamic acid tertbutyl ester, di-tert-butyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, boc-py
PubChem CID 11129853
IUPAC Name ditert-butyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C

Specifications

Melting Point 58°C
Color White
Quantity 5 g
Formula Weight 285.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl N-(tert-Butoxycarbonyl)-L-pyroglutamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.