accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 912444-00-9

IUPAC Name: 2-(2-methylpyrrolidin-2-yl)-1H-1,3-benzodiazole-4-carboxamide
Synonyms: 2-(2-methylpyrrolidin-2-yl)-1H-1,3-benzodiazole-4-carboxamide
Molecular Weight (g/mol): 244.3
Molecular Formula: C13H16N4O
InChi Key: JNAHVYVRKWKWKQ-UHFFFAOYNA-N
SMILES: CC1(CCCN1)C1=NC2=C(C=CC=C2N1)C(N)=O
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)