accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOH

CAS RN 91271-65-7

IUPAC Name: {4-[(morpholin-4-yl)methyl]phenyl}methanol
Synonyms: [4-(morpholin-4-ylmethyl)phenyl]methanol
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: MWVQMAWLNHACQK-UHFFFAOYSA-N
SMILES: OCC1=CC=C(CN2CCOCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results