accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91271-82-8

IUPAC Name: 4-{[2-(azaniumylmethyl)phenyl]methyl}morpholin-4-ium
Synonyms: 4-{[2-(ammoniomethyl)phenyl]methyl}morpholin-4-ium
Molecular Weight (g/mol): 208.3
Molecular Formula: C12H20N2O
InChi Key: NMFAEZHWSZZJOA-UHFFFAOYSA-P
SMILES: [NH3+]CC1=CC=CC=C1C[NH+]1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results