CAS RN 912850-81-8
CAS RN 912850-81-8
IUPAC Name:
9-benzoyl-3,6-dibromo-9H-carbazole
Synonyms:
9-benzoyl-3,6-dibromocarbazole
Molecular Weight (g/mol):
429.11
Molecular Formula:
C19H11Br2NO
InChi Key:
SQONRPMZFYXHTA-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)N(C(=O)C1=CC=CC=C1)C1=C2C=C(Br)C=C1
1
–
1
of
1
results
1
9-Benzoyl-3,6-dibromo-9H-carbazole 98.0+%, TCI America™
CAS: 912850-81-8 Molecular Formula: C19H11Br2NO Molecular Weight (g/mol): 429.111 InChI Key: SQONRPMZFYXHTA-UHFFFAOYSA-N PubChem CID: 132274866 IUPAC Name: (3,6-dibromocarbazol-9-yl)-phenylmethanone SMILES: C1=CC=C(C=C1)C(=O)N2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br