Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Benzoyl-3,6-dibromo-9H-carbazole 98.0+%, TCI America™
SDP

Catalog No. B54721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B54721G Supplier TCI America Supplier No. B54721G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 912850-81-8
Molecular Formula C19H11Br2NO
Molecular Weight (g/mol) 429.111
InChI Key SQONRPMZFYXHTA-UHFFFAOYSA-N
PubChem CID 132274866
IUPAC Name (3,6-dibromocarbazol-9-yl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)N2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br

Specifications

Melting Point 217°C
Color White
Quantity 1 g
Formula Weight 429.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzoyl-3,6-dibromo-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.