CAS RN 912921-26-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ 4-ethynyl-N-ethyl-1,8-naphthalimide
Fluorescent probe for imaging of cell surface, intracellular fucosylated glycoproteins and glycolipids