accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2

CAS RN 91298-74-7

IUPAC Name: 2-methoxy-1-phenylethan-1-amine
Synonyms: 2-methoxy-1-phenylethanamine
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: CMTDMIYJXVBUDX-UHFFFAOYNA-N
SMILES: COCC(N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results