Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Methoxy-1-phenylethylamine, 96%, Thermo Scientific™

Catalog No. AAH27404MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27404MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH27404MD Supplier Thermo Scientific Chemicals Supplier No. H27404MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 91298-74-7
Molecular Formula C9H13NO
Quantity 250mg
MDL Number MFCD01862273
UN Number UN2735
Synonym s-2-methoxy-1-phenylethanamine, s-+-1-amino-1-phenyl-2-methoxyethane, s-2-methoxy-1-phenylethan-1-amine, 1s-2-methoxy-1-phenylethanamine, s-1-phenyl-2-methoxyethanamine, 1s-2-methoxy-1-phenylethylamine, s-1-amino-1-phenyl-2-methoxyethane, 1s-2-methoxy-1-phenylethan-1-amine, s-+-2-methoxy-1-phenylethylamine, benzenemethanamine, alpha-methoxymethyl-, alphas
InChI Key CMTDMIYJXVBUDX-SECBINFHSA-N
SMILES COCC(C1=CC=CC=C1)N
IUPAC Name (1S)-2-methoxy-1-phenylethanamine
Molecular Weight (g/mol) 151.209
PubChem CID 11768756
Formula Weight 151.21
Percent Purity 96%
Chemical Name or Material (S)-(+)-2-Methoxy-1-phenylethylamine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.