CAS RN 91300-60-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ LY 83583
A cell-permeable, competitive inhibitor of soluble guanylate cyclase (IC50 = 2 μM).