Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ LY 83583

Catalog No. 4402055MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-020-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-020-55MG Supplier MilliporeSigma™ Supplier No. 4402055MG
Add to Cart
Add to Cart

A cell-permeable, competitive inhibitor of soluble guanylate cyclase (IC50 = 2 μM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Reddish Purple
Description 98% by TLC and HPLC
Target Proteins soluble guanylate cyclase
CAS 91300-60-6
Molecular Formula C15H10N2O2
Formula Weight 250.2g/mol
Solubility EtOH
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.