accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 913835-57-1

IUPAC Name: {3-[(2-hydroxyethyl)sulfamoyl]phenyl}boronic acid
Synonyms: 3-[(2-hydroxyethyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol): 245.06
Molecular Formula: C8H12BNO5S
InChi Key: HSOUGLHWSPWPOM-UHFFFAOYSA-N
SMILES: OCCNS(=O)(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results