accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONN

CAS RN 91419-53-3

IUPAC Name: tert-butyl 3-cyanopiperidine-1-carboxylate
Synonyms: tert-butyl 3-cyanopiperidine-1-carboxylate
Molecular Weight (g/mol): 210.28
Molecular Formula: C11H18N2O2
InChi Key: UEFZTXGFHKPSFS-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC(C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results