Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butoxycarbonyl-3-cyanopiperidine 98.0+%, TCI America™
SDP

Catalog No. B45155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B45155G Supplier TCI America Supplier No. B45155G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91419-53-3
Molecular Formula C11H18N2O2
Molecular Weight (g/mol) 210.277
MDL Number MFCD01861222
InChI Key UEFZTXGFHKPSFS-UHFFFAOYSA-N
Synonym 1-n-boc-3-cyanopiperidine, n-boc-3-cyanopiperidine, 1-boc-3-cyanopiperidine, +/--1-n-boc-3-cyano-piperidine, n-boc-3-cyano-piperidine, 1-n-boc-3-cyano-piperidine, 3-cyano-piperidine-1-carboxylic acid tert-butyl ester, 1-n-boc-piperidine-3-carbonitrile, 1-tert-butoxycarbonyl-3-cyanopiperidine, tert-butyl-3-cyanopiperidine-1-carboxylate
PubChem CID 2756785
IUPAC Name tert-butyl 3-cyanopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C#N

Specifications

Melting Point 67°C
Color White
Boiling Point 40°C
Quantity 5 g
UN Number 3439
Formula Weight 210.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butoxycarbonyl-3-cyanopiperidine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.