accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91421-42-0

IUPAC Name: (19R)-19-ethyl-19-hydroxy-8-nitro-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Synonyms: rubitecan
Molecular Weight (g/mol): 393.36
Molecular Formula: C20H15N3O6
InChi Key: VHXNKPBCCMUMSW-HXUWFJFHSA-N
SMILES: CC[C@]1(O)C(=O)OCC2=C1C=C1N(CC3=C1N=C1C=CC=C(C1=C3)[N+]([O-])=O)C2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results