Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Nitrocamptothecin 98.0+%, TCI America™
SDP

Catalog No. N0822100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0822100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0822100MG Supplier TCI America Supplier No. N0822100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91421-42-0
Molecular Formula C20H15N3O6
Molecular Weight (g/mol) 393.355
MDL Number MFCD06656294
InChI Key VHXNKPBCCMUMSW-FQEVSTJZSA-N
Synonym Rubitecan
PubChem CID 472335
SMILES CCC1(C2=C(COC1=O)C(=O)N3CC4=C(C3=C2)N=C5C=CC=C(C5=C4)[N+](=O)[O-])O

Specifications

Melting Point 268°C
Color Yellow
Quantity 100 mg
Formula Weight 393.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Nitrocamptothecin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.