accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91604-41-0

IUPAC Name: 1,4-bis(benzyloxy)butane-2,3-diol
Synonyms: 1,4-bis(benzyloxy)butane-2,3-diol
Molecular Weight (g/mol): 302.37
Molecular Formula: C18H22O4
InChi Key: YAVAVQDYJARRAU-UHFFFAOYNA-N
SMILES: OC(COCC1=CC=CC=C1)C(O)COCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results