Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-1,4-Di-O-benzyl-D-threitol 98.0+%, TCI America™
SDP

Catalog No. D22391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22391G Supplier TCI America Supplier No. D22391G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91604-41-0
Molecular Formula C18H22O4
Molecular Weight (g/mol) 302.37
MDL Number MFCD00077722
InChI Key YAVAVQDYJARRAU-UHFFFAOYNA-N
Synonym (+)-(2R,3R)-1,4-Bis(benzyloxy)-2,3-butanediol
PubChem CID 7004210
IUPAC Name 1,4-bis(benzyloxy)butane-2,3-diol
SMILES OC(COCC1=CC=CC=C1)C(O)COCC1=CC=CC=C1

Specifications

Melting Point 56°C
Color White-Yellow
Quantity 1 g
Formula Weight 302.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-1,4-Di-O-benzyl-D-threitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.