accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrICl

CAS RN 916203-52-6

IUPAC Name: 4-bromo-2-chloro-3-iodopyridine
Synonyms: 4-bromo-2-chloro-3-iodopyridine
Molecular Weight (g/mol): 318.34
Molecular Formula: C5H2BrClIN
InChi Key: ALKCOVKKTNNOGX-UHFFFAOYSA-N
SMILES: ClC1=NC=CC(Br)=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results