accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 917-64-6

IUPAC Name: iodo(methyl)magnesium
Synonyms: iodo(methyl)magnesium
Molecular Weight (g/mol): 166.24
Molecular Formula: CH3IMg
InChi Key: AUPXBVDHVRZMIB-UHFFFAOYSA-M
SMILES: C[Mg]I
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)