Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methylmagnesium Iodide Solution

Catalog No. 254363800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-436-3800ML 800 mL
25-436-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 25-436-3800ML Supplier Sigma Aldrich Supplier No. 254363800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methylmagnesium iodide solution can be used: To prepare cis-1,2-dialkylcyclopropanols by treating with titanium(IV) alkoxide and carboxylic esters. In stereospecific Ni catalyzed Kumada cross-coupling reactions of benzylic ethers. To prepare 2,4-disubstituted selenochromenes by reacting with 1-benzoselenopyrylium salts. In one of the key synthetic steps for the preparation of 7+a-methyl-substituted 5-androstene derivatives from 3+a-acetoxyandrost-5-en-17-one stereoselectively.

Specifications

CAS 917-64-6
Density 1.261 g/mL at 25°C
Molecular Formula CH3IMg
Linear Formula CH3MgI
Quantity 800 mL
MDL Number MFCD00001026
Molecular Weight (g/mol) 166.24 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.