accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91702-98-6

IUPAC Name: 3-(acetylsulfanyl)-2-benzylpropanoic acid
Synonyms: 3-(acetylsulfanyl)-2-benzylpropanoic acid
Molecular Weight (g/mol): 238.3
Molecular Formula: C12H14O3S
InChi Key: BCAAXVOKLXDSPD-UHFFFAOYNA-N
SMILES: CC(=O)SCC(CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results