Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[(Acetylthio)methyl]-3-phenylpropionic Acid 98.0+%, TCI America™
SDP

Catalog No. A22205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22205G 5 g
A22201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22205G Supplier TCI America Supplier No. A22205G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91702-98-6
Molecular Formula C12H14O3S
Molecular Weight (g/mol) 238.301
MDL Number MFCD00236751
InChI Key BCAAXVOKLXDSPD-UHFFFAOYSA-N
PubChem CID 9991796
IUPAC Name 2-(acetylsulfanylmethyl)-3-phenylpropanoic acid
SMILES CC(=O)SCC(CC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 39°C
Color White-Yellow
Quantity 5 g
Formula Weight 238.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-[(Acetylthio)methyl]-3-phenylpropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.