accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOH

CAS RN 91717-85-0

IUPAC Name: decane-1,2,10-triol
Synonyms: decane-1,2,10-triol
Molecular Weight (g/mol): 190.28
Molecular Formula: C10H22O3
InChi Key: RHINSRUDDXGHLV-UHFFFAOYNA-N
SMILES: OCCCCCCCCC(O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results