Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,10-Decanetriol 97.0+%, TCI America™
SDP

Catalog No. D27215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27215G Supplier TCI America Supplier No. D27215G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 91717-85-0
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.283
MDL Number MFCD00797600
InChI Key RHINSRUDDXGHLV-UHFFFAOYSA-N
Synonym 1,2,10-Trihydroxydecane
PubChem CID 2735912
IUPAC Name decane-1,2,10-triol
SMILES C(CCCCO)CCCC(CO)O

Specifications

Melting Point 66°C
Color White
Quantity 5 g
Formula Weight 190.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,10-Decanetriol
EINECSNumber (2)-2391
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.